Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

740489

Sigma-Aldrich

Trimethylsulfonium tetrafluoroborate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H9BF4S
CAS Number:
Molecular Weight:
163.97
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

reaction type: C-C Bond Formation

mp

205-210 °C

functional group

thioether

storage temp.

2-8°C

SMILES string

C[S+](C)C.F[B-](F)(F)F

InChI

1S/C3H9S.BF4/c1-4(2)3;2-1(3,4)5/h1-3H3;/q+1;-1

InChI key

RNBNWTAKWJKQNN-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service