Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

737534

Sigma-Aldrich

1-Z-2-Piperidone

95%

Synonym(s):

1-N-(Benzyloxycarbonyl)-2-piperidone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H15NO3
CAS Number:
Molecular Weight:
233.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.544

density

1.194 g/mL at 25 °C

functional group

phenyl

SMILES string

O=C1CCCCN1C(=O)OCc2ccccc2

InChI

1S/C13H15NO3/c15-12-8-4-5-9-14(12)13(16)17-10-11-6-2-1-3-7-11/h1-3,6-7H,4-5,8-10H2

InChI key

LKGMGCSFOUSKCR-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service