Skip to Content
MilliporeSigma
All Photos(2)

Documents

700223

Sigma-Aldrich

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)manganese(III)

97%

Synonym(s):

Mn(dpm)3, Shenvi Hydrogenation Catalyst, Mn(TMHD)3, Tris(dipivaloylmethanato)manganese

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mn(C11H19O2)3
CAS Number:
Molecular Weight:
604.74
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

crystals

reaction suitability

core: manganese
reagent type: catalyst

mp

160-170 °C

SMILES string

CC(C)(C)C(=O)\C=C(/O[Mn](O\C(=C/C(=O)C(C)(C)C)C(C)(C)C)O\C(=C/C(=O)C(C)(C)C)C(C)(C)C)C(C)(C)C

InChI

1S/3C11H20O2.Mn/c3*1-10(2,3)8(12)7-9(13)11(4,5)6;/h3*7,12H,1-6H3;/q;;;+3/p-3/b3*8-7-;

InChI key

ORZPLLPBZHORSD-LWTKGLMZSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for:
  • Intramolecular Diels-Alder reactions
  • Single electron donor for excess electron transfer studies in DNA
  • Enantioselective synthesis
  • Borylation reactions
  • Hydrohydrazination and hydroazidation
  • Oxidative carbonylation of phenol
Notably, this non-precious metal catalyst can be used to obtain the thermodynamic hydrogenation product of olefins, selectively.

Simple, Chemoselective Hydrogenation with Thermodynamic Stereocontrol

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service