Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

735736

Sigma-Aldrich

1-Boc-3-(aminomethyl)piperidine

90%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H22N2O2
CAS Number:
Molecular Weight:
214.30
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

90%

form

liquid

refractive index

n20/D 1.469

density

0.995 g/mL at 25 °C

functional group

amine

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)N1CCCC(CN)C1

InChI

1S/C11H22N2O2/c1-11(2,3)15-10(14)13-6-4-5-9(7-12)8-13/h9H,4-8,12H2,1-3H3

InChI key

WPWXYQIMXTUMJB-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service