Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

731560

Sigma-Aldrich

3-Iodooxetane

95%

Synonym(s):

1,3-Epoxy-2-iodopropane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5IO
CAS Number:
Molecular Weight:
183.98
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

availability

not available in EU

refractive index

n20/D 1.563

density

2.090 g/mL at 25 °C

functional group

ether
iodo

storage temp.

2-8°C

SMILES string

IC1COC1

InChI

1S/C3H5IO/c4-3-1-5-2-3/h3H,1-2H2

InChI key

KBEIFKMKVCDETC-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

150.1 °F

Flash Point(C)

65.6 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service