Skip to Content
MilliporeSigma
All Photos(1)

Documents

722456

Sigma-Aldrich

4-Bromotetrahydropyran

Synonym(s):

4-Bromooxacyclohexane, 4-Bromotetrahydro-2H-pyran

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9BrO
CAS Number:
Molecular Weight:
165.03
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

refractive index

n20/D 1.497

density

1.467 g/cm3 at 25 °C

SMILES string

BrC1CCOCC1

InChI

1S/C5H9BrO/c6-5-1-3-7-4-2-5/h5H,1-4H2

InChI key

IVBVKTPDEWDNRW-UHFFFAOYSA-N

Application

Reactant for:
  • Nickel-catalyzed alkyl-alkyl Suzuki coupling reactions with boron reagents
  • Preparation of a selective small-molecule melanocortin-4 receptor agonist with efficacy in a pilot study of sexual dysfunction in humans
  • Preparation of aliphatic hydrocarbons via nickel-catalyzed Suzuki cross-coupling with alkylboranes
  • Preparation of anthranilic acids as antibacterial agents with human serum albumin binding affinity
  • Preparation of antiatherogenic antioxidant di-tert-butyldihydrobenzofuranols via Grignard reactions with di-tert-butyl(hydroxy)benzaldehyde derivatives
  • Synthesis of gephyrotoxin via the Schmidt reaction

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

160.0 °F - closed cup

Flash Point(C)

71.1 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service