Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

731358

Sigma-Aldrich

8-Bromo-3-methoxyisoquinoline

≥90%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8BrNO
CAS Number:
Molecular Weight:
238.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥90%

form

solid

mp

61-67 °C

functional group

bromo

SMILES string

COc1cc2cccc(Br)c2cn1

InChI

1S/C10H8BrNO/c1-13-10-5-7-3-2-4-9(11)8(7)6-12-10/h2-6H,1H3

InChI key

PKOWJHRDTZPIHB-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service