Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3623

Sigma-Aldrich

DSRM-3716

≥98% (HPLC)

Synonym(s):

5-Iodoisoquinoline, DSRM 3716, DSRM3716

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6IN
CAS Number:
Molecular Weight:
255.06
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

, yellow to light brown

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

Biochem/physiol Actions

DSRM-3716 is a potent and selective SARM1 NADase inhibitor (NAD+ hydrolysis IC50 = 75 nM using recombinant SARM1 SAM-TIR construct) that recapitulates the SARM1-/- phenotype and protects axons from degeneration induced by axotomy (EC50 = 2.1/1.9 µM by fragmentation/NfL release using mouse DRG neurons; 81/88% protection by 1/3 µM DSRM-3716 using human iPSC-derived motor neurons) or mitochondrial dysfunction (3-30 µM DSRM-3716 against 25 µM rotenone-induced injury; mouse DRG neurons). Mechanistically, DSRM-3716 undergoes base exchange with the nicotinamide (NAM) moiety of nicotinamide adenine dinucleotide (NAD+) to yield the bona fide inhibitor 1AD.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural basis of SARM1 activation, substrate recognition, and inhibition by small molecules.
Shi Y, et al.
Molecular Cell, 82, 1643-1659 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service