Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

724963

Sigma-Aldrich

Methanethiol-13C

99 atom % 13C, 97% (CP)

Synonym(s):

Methyl-13C mercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CH3SH
CAS Number:
Molecular Weight:
49.10
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

97% (CP)

form

gas

bp

6 °C (lit.)

mass shift

M+1

SMILES string

[13CH3]S

InChI

1S/CH4S/c1-2/h2H,1H3/i1+1

InChI key

LSDPWZHWYPCBBB-OUBTZVSYSA-N

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Gas 1A - Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

-0.4 °F - closed cup

Flash Point(C)

-18 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service