Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

724041

Sigma-Aldrich

NG,NG-Dimethyl-d6-L-arginine dihydrochloride

98 atom % D, 95% (CP)

Synonym(s):

asym-Dimethyl-d6-arginine, ADMA-d6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8D6H14Cl2N4O2
Molecular Weight:
281.21
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

95% (CP)

form

powder

technique(s)

bio NMR: suitable

mass shift

M+6

storage temp.

−20°C

SMILES string

Cl.Cl.[2H]C([2H])([2H])N(C(=N)NCCC[C@H](N)C(O)=O)C([2H])([2H])[2H]

InChI

1S/C8H18N4O2.2ClH/c1-12(2)8(10)11-5-3-4-6(9)7(13)14;;/h6H,3-5,9H2,1-2H3,(H2,10,11)(H,13,14);2*1H/t6-;;/m0../s1/i1D3,2D3;;

InChI key

SYLNVYJOPZWPJI-WTPXGJEESA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service