Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

678406

Sigma-Aldrich

Methyl jasmonate-(methyl acetate-2,2-d2), racemic

97 atom % D, 95% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13D2H18O3
CAS Number:
Molecular Weight:
226.31
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

isotopic purity

97 atom % D

Assay

95% (CP)

bp

110 °C/0.2 mmHg

density

1.039 g/mL at 25 °C

mass shift

M+2

SMILES string

[2H]C([2H])(C1CCC(=O)C1C\C=C/CC)C(=O)OC

InChI

1S/C13H20O3/c1-3-4-5-6-11-10(7-8-12(11)14)9-13(15)16-2/h4-5,10-11H,3,6-9H2,1-2H3/b5-4-/i9D2

InChI key

GEWDNTWNSAZUDX-GZSAMHGASA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F

Flash Point(C)

113 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service