Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

717274

Sigma-Aldrich

1,3,6,8-Tetrabromopyrene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H6Br4
CAS Number:
Molecular Weight:
517.83
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

mp

416-420 °C

SMILES string

Brc1cc(Br)c2ccc3c(Br)cc(Br)c4ccc1c2c34

InChI

1S/C16H6Br4/c17-11-5-13(19)9-3-4-10-14(20)6-12(18)8-2-1-7(11)15(9)16(8)10/h1-6H

InChI key

ZKBKRTZIYOKNRG-UHFFFAOYSA-N

Application

Synthetic building block for the creation of blue to green OLED emitters

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasuchika Hasegawa et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(53), 12308-12315 (2019-07-23)
Oxygen-sensitive and near-infrared (NIR) luminescent YbIII coordination polymers incorporating ligands based on pyrene derivatives were synthesized: YbIII -TBAPy and YbIII -TIAPy (TBAPy: 1,3,6,8-tetrakis(p-benzoate)pyrene; TIAPy: 1,3,6,8-tetrakis(3,5-isophthalic acid)pyrene). The coordination structures of these materials have been characterized by means of electrospray ionization
You Zhou et al.
The Analyst, 143(15), 3628-3634 (2018-07-04)
1-Hydroxypyrene (1-HP) is a urinary metabolite of polycyclic aromatic hydrocarbons (PAHs), and can function as a convenient biomarker for human intoxication of PAH carcinogens. The development of simple 1-HP sensors with high sensitivity and fast response is highly desirable. Herein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service