Skip to Content
MilliporeSigma
All Photos(2)

Documents

715670

Sigma-Aldrich

2-(Bromoacetyl)benzofuran

97%

Synonym(s):

1-(1-Benzofuran-2-yl)-2-bromoethan-1-one, 1-(1-Benzofuran-2-yl)-2-bromoethanone, 1-(2-Benzofuryl)-2-bromo-1-ethanone, 1-(Benzo[b]furan-2-yl)-2-bromoethan-1-one, 1-(Benzofuran-2-yl)-2-bromoethanone, 1-Benzo[b]furan-2-yl-2-bromoethanone, 2-(2-Bromoacetyl)benzofuran, 2-(Bromoacetyl)benzo[b]furan, 2-Benzofuranyl bromomethyl ketone, 2-Benzofuryl bromomethyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7BrO2
CAS Number:
Molecular Weight:
239.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

86-90 °C

SMILES string

BrCC(=O)c1cc2ccccc2o1

InChI

1S/C10H7BrO2/c11-6-8(12)10-5-7-3-1-2-4-9(7)13-10/h1-5H,6H2

InChI key

NVRNCBWTEDOAQA-UHFFFAOYSA-N

Application

Reactant for:
  • Eschenmoser coupling reaction
  • Preparation of β-oxo sulfones as substrates for stereoselective Michael addition/cross-benzoin cascade reactions
  • Preparation of thienylpyrazole-based thiazoles and pyrazolines as antimicrobial, antioxidant, anti-inflammatory, and analgesic agents
  • Preparation of isatin-(benzofuryl-thiazolyl)-hydrazones by cyclocondensation with isatin-thiosemicarbazones under microwave irradiation as antimicrobial agents
  • Synthesis of 2-naphthyl ethers as a protective agent against DNA damage induced by bleomycin-iron
  • Asymmetric synthesis of β-dialkylamino alcohols by Ru-catalyzed transfer hydrogenation of α-dialkylamino ketones

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Veeraiah
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 147, 212-224 (2015-04-05)
The vibrational and electronic properties of 2-(bromoacetyl)benzo(b)furan have been studied in the ground state using experimental techniques (FT-IR, FT-Raman and UV) and density functional theory (DFT) employing B3LYP exchange correlation with the 6-31G(d,p) basis set. The theoretically calculated optimized parameters
Justin D Smith et al.
Nature communications, 10(1), 1837-1837 (2019-04-25)
Photocatalytic polymers offer an alternative to prevailing organometallics and nanomaterials, and they may benefit from polymer-mediated catalytic and material enhancements. MPC-1, a polymer photoredox catalyst reported herein, exhibits enhanced catalytic activity arising from charge transfer states (CTSs) between its two

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service