Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

224340

Sigma-Aldrich

2-Methylbenzofuran

96%

Synonym(s):

2-Methyl-1-benzofuran, 2-Methylbenzo[b]furan, 2-Methylcumarone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8O
CAS Number:
Molecular Weight:
132.16
Beilstein:
2165
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

refractive index

n20/D 1.560 (lit.)

bp

197-198 °C (lit.)

density

1.057 g/mL at 25 °C (lit.)

SMILES string

Cc1cc2ccccc2o1

InChI

1S/C9H8O/c1-7-6-8-4-2-3-5-9(8)10-7/h2-6H,1H3

InChI key

GBGPVUAOTCNZPT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methylbenzofuran, also known as 2-Methylcumarone is a chemical intermediate and building block used for synthesizing various organic compounds. It also acts as a substrate in Friedel-Crafts alkylation and acylation reactions.

Application

2-Methylbenzofuran can be used as a reactant to synthesize enol derivatives via catalytic oxidation by hydrogen peroxide.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

152.6 °F - closed cup

Flash Point(C)

67 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biomimetic Oxidation of Benzofurans with Hydrogen Peroxide Catalyzed by Mn (III) Porphyrins
Rebelo S, et al.
Catalysts, 10, 62-62 (2020)
Xichao Chen et al.
Journal of environmental sciences (China), 25(11), 2313-2323 (2014-02-21)
Taste and odor (T&O) problems in drinking water frequently occur because of many compounds present in the water, of which trans-1,10-dimethyl-trans-9-decalol (geosmin) and 2-methylisoborneol (MIB) are well-known. In this study, a fast and effective method was established for simultaneous determination

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service