Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

715271

Sigma-Aldrich

3,5-Dimethoxyphenylboronic acid

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11BO4
CAS Number:
Molecular Weight:
181.98
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95%

form

solid

mp

202-207 °C

SMILES string

COc1cc(OC)cc(c1)B(O)O

InChI

1S/C8H11BO4/c1-12-7-3-6(9(10)11)4-8(5-7)13-2/h3-5,10-11H,1-2H3

InChI key

XUIURRYWQBBCCK-UHFFFAOYSA-N

Application

Reactant for:
  • Palladium-catalyzed coupling reactions
  • Preparation of benzopyranone derivatives as positive GABAA receptor modulators
  • Preparation of aryl alkenes via three-component coupling catalyzed by palladium
  • Suzuki-Miyaura coupling
  • Rhodium catalyzed cyanation with N-cyano-N-phenyl-p-methylbenzenesulfonamide
  • Preparation of bisphosphonate inhibitors of human farnesyl pyrophosphate synthase

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

178.3 °F - closed cup

Flash Point(C)

81.3 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service