Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

542318

Sigma-Aldrich

2,4,6-Trimethylphenylboronic acid

≥95%

Synonym(s):

2,4,6-Trimethylbenzeneboronic acid, Mesitylene-2-boronic acid, NSC 157832

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H2(CH3)3B(OH)2
CAS Number:
Molecular Weight:
164.01
Beilstein:
2831927
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder

mp

115-122 °C (lit.)

SMILES string

Cc1cc(C)c(B(O)O)c(C)c1

InChI

1S/C9H13BO2/c1-6-4-7(2)9(10(11)12)8(3)5-6/h4-5,11-12H,1-3H3

InChI key

BZXQRXJJJUZZAJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Contains varying amounts of anhydride.

Application

Reactant involved in:
  • Cross-coupling with α-bromocarbonyl compounds or 2,6-disubstituted bromoarenes
  • Suzuki-Miyaura coupling for synthesis of biphenyl arsine ligands
  • Addition reactions to naphthyridine N-oxides or β-aryloxyacrylates

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ken T Ngo et al.
Journal of the American Chemical Society, 139(7), 2604-2618 (2017-01-25)
Electrocatalytic reduction of CO

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service