Skip to Content
MilliporeSigma
All Photos(1)

Documents

704180

Sigma-Aldrich

DL-2,4-Diaminobutyric-3,3,4,4-d4 acid dihydrochloride

98 atom % D, 98% (CP)

Synonym(s):

DL-2,4-Diaminobutanoic-3,3,4,4-d4 acid dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CD2CD2CH(NH2)COOH .2HCl
CAS Number:
Molecular Weight:
195.08
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

98% (CP)

form

powder

mol wt

195.01 by atom % calculation

technique(s)

mass spectrometry (MS): suitable

mp

197 °C (dec.)

mass shift

M+4

SMILES string

Cl.Cl.[2H]C([2H])(N)C([2H])([2H])C(N)C(O)=O

InChI

1S/C4H10N2O2.2ClH/c5-2-1-3(6)4(7)8;;/h3H,1-2,5-6H2,(H,7,8);2*1H/i1D2,2D2;;

InChI key

CKAAWCHIBBNLOJ-RIZDZYNXSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service