Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

702242

Sigma-Aldrich

Sodium pyruvate-2-13C, 3,3,3-d3

99 atom % 13C, 97 atom % D, 98% (CP)

Synonym(s):

Pyruvic acid-2-13C,3,3,3-d3 sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD313COCO2Na
Molecular Weight:
114.06
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:

isotopic purity

99 atom % 13C
97 atom % D

Assay

98% (CP)

form

powder

mol wt

113.96 by atom % calculation

technique(s)

mass spectrometry (MS): suitable

mp

>300 °C

mass shift

M+4

storage temp.

2-8°C

SMILES string

[Na+].[2H]C([2H])([2H])[13C](=O)C([O-])=O

InChI

1S/C3H4O3.Na/c1-2(4)3(5)6;/h1H3,(H,5,6);/q;+1/p-1/i1D3,2+1;

InChI key

DAEPDZWVDSPTHF-CKHMYQJJSA-M

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service