Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

702005

Sigma-Aldrich

1,1′-Bis(di-isopropylphosphino)ferrocene palladium dichloride

98%

Synonym(s):

PdCl2(dippf)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H36Cl2FeP2Pd
CAS Number:
Molecular Weight:
595.64
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

powder

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

282-287 °C

storage temp.

−20°C

SMILES string

[Fe].Cl[Pd]Cl.CC(C)P([C]1[CH][CH][CH][CH]1)C(C)C.CC(C)P([C]2[CH][CH][CH][CH]2)C(C)C

InChI

1S/2C11H18P.2ClH.Fe.Pd/c2*1-9(2)12(10(3)4)11-7-5-6-8-11;;;;/h2*5-10H,1-4H3;2*1H;;/q;;;;;+2/p-2

InChI key

UXOSTJATWDHIKQ-UHFFFAOYSA-L

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service