Skip to Content
MilliporeSigma
All Photos(3)

Documents

675784

Sigma-Aldrich

1,2,3,4,5-Pentaphenyl-1′-(di-tert-butylphosphino)ferrocene

Synonym(s):

QPhos

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(C6H5)5C5]Fe[C5H4(P(C4H9)2)]
CAS Number:
Molecular Weight:
710.71
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reagent type: ligand
reaction type: Arylations

reagent type: ligand
reaction type: Buchwald-Hartwig Cross Coupling Reaction

reagent type: ligand
reaction type: Sonogashira Coupling

reagent type: ligand
reaction type: Suzuki-Miyaura Coupling

mp

211-219 °C

functional group

phosphine

SMILES string

[Fe].CC(C)(C)P([C]1[CH][CH][CH][CH]1)C(C)(C)C.c2ccc(cc2)[C]3[C]([C]([C]([C]3c4ccccc4)c5ccccc5)c6ccccc6)c7ccccc7

InChI

1S/C35H25.C13H22P.Fe/c1-6-16-26(17-7-1)31-32(27-18-8-2-9-19-27)34(29-22-12-4-13-23-29)35(30-24-14-5-15-25-30)33(31)28-20-10-3-11-21-28;1-12(2,3)14(13(4,5)6)11-9-7-8-10-11;/h1-25H;7-10H,1-6H3;

InChI key

YOQRDWPCPFCDLP-UHFFFAOYSA-N

Application

Sterically hindered phosphine with broad application in a variety of palladium-catalyzed amination and etherification reactions of aryl chlorides.

Legal Information

Sold in collaboration with Johnson Matthey Catalyst for research purposes only. US6562989 B2 and any patents arising therefrom apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takuo Hama et al.
Journal of the American Chemical Society, 128(15), 4976-4985 (2006-04-13)
The intermolecular alpha-arylation and vinylation of amides by palladium-catalyzed coupling of aryl bromides and vinyl bromides with zinc enolates of amides is reported. Reactions of three different types of zinc enolates have been developed. The reactions of aryl halides occur

Articles

Q-Phos ligand by Professor Hartwig enables diverse C-N, C-O, and C-C bondforming reactions in coupling reactions.

Q-Phos ligand by Professor Hartwig enables diverse C-N, C-O, and C-C bondforming reactions in coupling reactions.

Q-Phos ligand by Professor Hartwig enables diverse C-N, C-O, and C-C bondforming reactions in coupling reactions.

Q-Phos ligand by Professor Hartwig enables diverse C-N, C-O, and C-C bondforming reactions in coupling reactions.

Related Content

Professor Hartwig's research focuses on the discovery and understanding of new reactions catalyzed by transition metal complexes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service