Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

700517

Sigma-Aldrich

4-Nitro-α,α,α-trifluorotoluene-(phenyl-13C6)

99 atom % 13C, 97% (CP)

Synonym(s):

4-Nitrobenzotrifluoride-13C6, (phenyl-13C6)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13C6F3H4NO2
Molecular Weight:
197.06
MDL number:
UNSPSC Code:
12352102
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

97% (CP)

form

solid

mp

38-40  °C

mass shift

M+6

SMILES string

[O-][N+](=O)[13c]1[13cH][13cH][13c]([13cH][13cH]1)C(F)(F)F

InChI

1S/C7H4F3NO2/c8-7(9,10)5-1-3-6(4-2-5)11(12)13/h1-4H/i1+1,2+1,3+1,4+1,5+1,6+1

InChI key

XKYLCLMYQDFGKO-IDEBNGHGSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service