Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

697451

Sigma-Aldrich

Dimethyl-d6 carbonate

99 atom % D, 97% (CP)

Synonym(s):

1,1′-carbonate methan-d3-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CD3O)2CO
CAS Number:
Molecular Weight:
96.11
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % D

Quality Level

Assay

97% (CP)

form

liquid

mol wt

96.06 (atom % calculation)

bp

90 °C

mp

2-4 °C

application(s)

pharmaceutical

mass shift

M+6

SMILES string

[2H]C([2H])([2H])OC(=O)OC([2H])([2H])[2H]

InChI

1S/C3H6O3/c1-5-3(4)6-2/h1-2H3/i1D3,2D3

InChI key

IEJIGPNLZYLLBP-WFGJKAKNSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

60.8 °F

Flash Point(C)

16 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service