Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

690791

Sigma-Aldrich

α-Methyl-D-phenylalanine

≥98.0% (HPLC)

Synonym(s):

α-Me-D-Phe-OH, (R)-(+)-2-Amino-2-methyl-3-phenylpropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13NO2
CAS Number:
Molecular Weight:
179.22
Beilstein:
5254151
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (HPLC)

form

powder

optical purity

ee: ≥98.0% (HPLC)

reaction suitability

reaction type: solution phase peptide synthesis

application(s)

peptide synthesis

SMILES string

C[C@@](N)(Cc1ccccc1)C(O)=O

InChI

1S/C10H13NO2/c1-10(11,9(12)13)7-8-5-3-2-4-6-8/h2-6H,7,11H2,1H3,(H,12,13)/t10-/m1/s1

InChI key

HYOWVAAEQCNGLE-SNVBAGLBSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O Greengard et al.
Biochemical pharmacology, 36(6), 965-970 (1987-03-15)
Severe hyperphenylalaninemia induced in infant rats by 3 days of treatment with p-chlorophenylalanine (p-cl phe) plus phenylalanine (phe) did not lower the tryptophan concentration of the brain, and the cerebral serotonin (5-HT) deficiency was attributable entirely to the known suppression
Shinya Oishi et al.
Journal of medicinal chemistry, 48(3), 764-772 (2005-02-04)
Previous work has shown that incorporation of either 1-aminocyclohexanecarboxylic acid (Ac6c) or alpha-methyl-p-phosphonophenylalanine ((alpha-Me)Ppp) in the phosphotyrosyl (pTyr) C-proximal position (pY + 1 residue) of Grb2 SH2 domain binding peptides confers high affinity. The tetralin-based (S)-2-amino-6-phosphonotetralin-2-carboxylic acid (Atc(6-PO3H2)) simultaneously presents
O Greengard et al.
Neurochemical research, 9(6), 837-848 (1984-06-01)
Brain phenylalanine concentrations at plasma levels raised to that in phenylketonuric subjects were studied in rats from fetal through postnatal life. Suppression of the hepatic phenylalanine hydroxylase with alpha methylphenylalanine, and injections of age-adjusted doses of phenylalanine on the next
C A Brass et al.
The Biochemical journal, 208(3), 765-771 (1982-12-15)
Hyperphenylalaninaemia induced by daily injections of alpha-methylphenylalanine plus phenylalanine caused 20-40% decreases in cerebral dopamine (3,4-dihydroxyphenethylamine) and noradrenaline in 7- and 11-day-old rats. alpha-Methylphenylalanine alone as well as phenylalanine alone caused cerebral dopamine depletion. However, the effects were not additive
Adverse effects of excessive consumption of amino acids.
N J Benevenga et al.
Annual review of nutrition, 4, 157-181 (1984-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service