Skip to Content
MilliporeSigma
All Photos(3)

Documents

392766

Sigma-Aldrich

AD-mix-β

Synonym(s):

AD-mix-beta

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

powder

Quality Level

SMILES string

CC[C@@H]1CN2CCC1CC2[C@H](Oc3nnc(O[C@@H](C4CC5CCN4C[C@@H]5CC)c6ccnc7ccc(OC)cc67)c8ccccc38)c9ccnc%10ccc(OC)cc9%10

InChI

1S/C48H54N6O4/c1-5-29-27-53-21-17-31(29)23-43(53)45(35-15-19-49-41-13-11-33(55-3)25-39(35)41)57-47-37-9-7-8-10-38(37)48(52-51-47)58-46(44-24-32-18-22-54(44)28-30(32)6-2)36-16-20-50-42-14-12-34(56-4)26-40(36)42/h7-16,19-20,25-26,29-32,43-46H,5-6,17-18,21-24,27-28H2,1-4H3/t29-,30-,31-,32-,43+,44+,45-,46-/m0/s1

InChI key

YUCBLVFHJWOYDN-HVLQGHBFSA-N

General description

Contains chiral ligand (DHQD)2PHAL

AD-mix-β is an oxidant used for the enantioselective synthesis of 2-Hydroxy-5-methyl-3-hexanone.

Application

Catalyst employed in the Sharpless Asymmetric Dihyroxylation of (E,E)- or (E,Z)-1,3-dienoates. Also used in the preparation of ß-hydroxy-γ-lactones from ß,γ-unsaturated esters.
Reagent for Sharpless Asymmetric Dihydroxylation.

Components

Mixture contains:
(DHQD)2PHAL (Cat. No. 392731) 0.0016 mole
Potassium carbonate, powder0.4988 mole
Potassium ferricyanide0.4988 mole
Potassium osmate dihydrate0.0007 mole

Legal Information

Sold under license from Rhodia Pharma Solutions.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of nonracemic 3-deoxyschweinfurthin B
Neighbors, Jeffrey D, et al.
The Journal of Organic Chemistry, 70, 925-931 (2005)
European Journal of Organic Chemistry, 2669-2669 (2006)
(4R)-4-[(1R)-1, 2-Dihydroxyethyl]-1-[(1R)-1-phenylethyl] pyrrolidin-2-one.
Blaser A and Boyd PDW.
Acta Crystallographica Section E, Structure Reports Online, 64(9), o1851-o1851 (2008)
Chemical Reviews, 94, 2483-2483 (1994)
First asymmetric synthesis of CJ-14877 and its enantiomer and their interleukin-1? inhibitory activities.
Aoyagi Y, et al.
Bioorganic & Medicinal Chemistry Letters, 19(7), 1876-1878 (2009)

Related Content

The Sharpless Lab pursues useful new reactivity and general methods for selectively controlling chemical reactions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service