Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

690082

Sigma-Aldrich

Ethyl benzoylacetate

Arxada quality, ≥91.0% (GC)

Synonym(s):

Benzoylacetic acid ethyl ester, Ethyl 3-oxo-3-phenylpropionate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COCH2COOC2H5
CAS Number:
Molecular Weight:
192.21
Beilstein:
389944
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

6.6 (vs air)

Assay

≥91.0% (GC)

form

liquid

quality

Arxada quality

manufacturer/tradename

Arxada AG

refractive index

n20/D 1.52 (lit.)

bp

265-270 °C (lit.)

density

1.11 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CC(=O)c1ccccc1

InChI

1S/C11H12O3/c1-2-14-11(13)8-10(12)9-6-4-3-5-7-9/h3-7H,2,8H2,1H3

InChI key

GKKZMYDNDDMXSE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

284.0 °F - closed cup

Flash Point(C)

140 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service