Skip to Content
MilliporeSigma
All Photos(1)

Documents

722464

Sigma-Aldrich

Ethyl-3-cyclopropyl-3-oxopropionate

96%

Synonym(s):

3-Cyclopropyl-3-oxo-propionic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12O3
CAS Number:
Molecular Weight:
156.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

liquid

refractive index

n20/D 1.449

density

1.058 g/mL at 25 °C

SMILES string

CCOC(=O)CC(=O)C1CC1

InChI

1S/C8H12O3/c1-2-11-8(10)5-7(9)6-3-4-6/h6H,2-5H2,1H3

InChI key

LFSVADABIDBSBV-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service