Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

690074

Sigma-Aldrich

2-Iodonaphthalene

≥99.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7I
CAS Number:
Molecular Weight:
254.07
Beilstein:
1905951
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (HPLC)

form

chunks

mp

52-56 °C

functional group

iodo

SMILES string

Ic1ccc2ccccc2c1

InChI

1S/C10H7I/c11-10-6-5-8-3-1-2-4-9(8)7-10/h1-7H

InChI key

FRNLBIWVMVNNAZ-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Long Zhao et al.
Physical chemistry chemical physics : PCCP, 21(30), 16737-16750 (2019-07-20)
The three-ring polycyclic aromatic hydrocarbons (PAHs) 3H-benz[e]indene (C13H10) and 1H-benz[f]indene (C13H10) along with their naphthalene-based isomers 2-(prop-2-yn-1-yl)naphthalene (C13H10), 2-(prop-1-yn-1-yl)naphthalene (C13H10), and 2-(propa-1,2-dien-1-yl)naphthalene (C13H10) were formed through a "directed synthesis"via a high temperature chemical micro reactor under combustion-like conditions (1300 ±

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service