Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

682411

Sigma-Aldrich

Di-tert-butylphenylphosphine

95%

Synonym(s):

NSC 244300

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H23P
CAS Number:
Molecular Weight:
222.31
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
reaction type: Cross Couplings

refractive index

n20/D 1.5382

bp

124-128 °C/10 mmHg

density

0.933 g/mL at 25 °C

functional group

phosphine

SMILES string

CC(C)(C)P(c1ccccc1)C(C)(C)C

InChI

1S/C14H23P/c1-13(2,3)15(14(4,5)6)12-10-8-7-9-11-12/h7-11H,1-6H3

InChI key

XOJNEFQLMRCOMS-UHFFFAOYSA-N

Application

Catalyst for:
  • Cross-coupling reactions
  • Palladium-catalyzed monoarylation reactions (cocatalyst)
  • Coupling of alkyl bromides with aryl boronic acids

  • Reactant for the synthesis of palladium-FiberCats for aryl chloride Suzuki coupling

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Pyr. Liq. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service