Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

68240

Sigma-Aldrich

Cetyl alcohol - stearyl alcohol mixture

Synonym(s):

Alcohol cetylis et stearylicus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352100
NACRES:
NA.22

Agency

USP/NF
tested according to Ph. Eur.

Related Categories

General description

Cetearyl alcohol is a combination of cetyl and stearyl fatty alcohols. It acts as an emulsion stabilizer, foam stabilizer, opacifying agent, and viscosity-increasing ingredient in both aqueous and nonaqueous solutions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simple and rapid analytical method for the simultaneous determination of cetrimonium chloride and alkyl alcohols in hair conditioners
Terol A, et al.
International Journal of Cosmetic Science, 32(1), 65-72 (2010)
Transdermal delivery of Nigella sativa oil for topical application in 1-chloro-2, 4-dinitrobenzene-induced atopic dermatitis in rats
Chawla R and Chauhan A
Journal of PharmaSciTech, 6, 51-59 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service