Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

258741

Sigma-Aldrich

1-Hexadecanol

ReagentPlus®, 99%

Synonym(s):

Cetyl alcohol, Palmityl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15OH
CAS Number:
Molecular Weight:
242.44
Beilstein:
1748475
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
grade:
reagent
bp:
179-181 °C/10 mmHg (lit.)
vapor pressure:
<0.01 mmHg ( 43 °C)
Pricing and availability is not currently available.

grade

reagent

vapor density

8.34 (vs air)

vapor pressure

<0.01 mmHg ( 43 °C)

product line

ReagentPlus®

Assay

99%

form

solid

autoignition temp.

483 °F

expl. lim.

8 %

dilution

(for general lab use)

bp

179-181 °C/10 mmHg (lit.)

mp

48-50 °C (lit.)

solubility

ethanol: soluble 100 mL/mL, clear to very slightly hazy, colorless

density

0.818 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CCCCCCCCCCCCCCCCO

InChI

1S/C16H34O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17/h17H,2-16H2,1H3

InChI key

BXWNKGSJHAJOGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Hexadecanol is a fatty alcohol used in surfactants, lubricants, detergents, pharmaceuticals and cosmetics. [1][2]

Application

1-Hexadecanol has been used in preparation of:
  • (±)-2-methoxyheptadecanoic acid (fatty acid)[3]
  • high-chain fatty acid esters of 1-hexadecanol, novel organic phase change material for thermal energy storage[4]
  • hexadecane (alkane) in the presence of membrane fraction of Vibrio furnissii M1[5]

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metabolic engineering of Saccharomyces cerevisiae to improve 1-hexadecanol production
Feng, et al.
Metabolic engineering, 27, 10-19 (2015)
Néstor M Carballeira et al.
Pure and applied chemistry. Chimie pure et appliquee, 84(9), 1867-1875 (2012-11-03)
The fatty acids (±)-2-methoxy-6Z-heptadecenoic acid (1), (±)-2-methoxy-6-heptadecynoic acid (2) and (±)-2-methoxyheptadecanoic acid (3) were synthesized and their inhibitory activity against the Leishmania DNA topoisomerase IB enzyme (LdTopIB) determined. Acids 1 and 2 were synthesized from 4-bromo-1-pentanol, the former in ten
Metabolic engineering of Saccharomyces cerevisiae to produce 1-hexadecanol from xylose
Guo, et al.
Microbial cell factories, 15, 1-11 (2016)
High-chain fatty acid esters of 1-hexadecanol for low temperature thermal energy storage with phase change materials.
Aydin AA and Aydin A.
Solar Energy Mat. and Solar Cells, 96, 93-100 (2012)
S Karen Gomez et al.
BMC plant biology, 9, 10-10 (2009-01-24)
Most vascular flowering plants have the capacity to form symbiotic associations with arbuscular mycorrhizal (AM) fungi. The symbiosis develops in the roots where AM fungi colonize the root cortex and form arbuscules within the cortical cells. Arbuscules are enveloped in

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service