Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

678759

Sigma-Aldrich

4-Iodo-2-(trifluoromethyl)benzonitrile

97%

Synonym(s):

4-Cyano-3-(trifluoromethyl)iodobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H3F3IN
CAS Number:
Molecular Weight:
297.02
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

50-55 °C

functional group

fluoro
iodo
nitrile

SMILES string

FC(F)(F)c1cc(I)ccc1C#N

InChI

1S/C8H3F3IN/c9-8(10,11)7-3-6(12)2-1-5(7)4-13/h1-3H

InChI key

RBRTXKYFMXVIER-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service