Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

677914

Sigma-Aldrich

2,4-Dibromothiazole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3HBr2NS
CAS Number:
Molecular Weight:
242.92
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

80-84 °C

functional group

bromo

SMILES string

Brc1csc(Br)n1

InChI

1S/C3HBr2NS/c4-2-1-7-3(5)6-2/h1H

InChI key

MKEJZKKVVUZXIS-UHFFFAOYSA-N

Application

Building block in a synthesis of Melithiazole C employing a highly (E)-selective cross-metathesis between the derived 4-vinylthiazole and a dienote side-chain.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julian Gebauer et al.
Organic letters, 9(17), 3425-3427 (2007-07-20)
A short and convergent synthesis of the myxobacterial antibiotic melithiazole C is described featuring a highly E-selective cross-metathesis as the key step.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service