Skip to Content
MilliporeSigma
All Photos(1)

Documents

642517

Sigma-Aldrich

5-Bromothiazole

95%

Synonym(s):

5-Bromo-1,3-thiazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H2BrNS
CAS Number:
Molecular Weight:
164.02
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

refractive index

n20/D 1.5955

density

1.835 at 25 °C

SMILES string

Brc1cncs1

InChI

1S/C3H2BrNS/c4-3-1-5-2-6-3/h1-2H

InChI key

DWUPYMSVAPQXMS-UHFFFAOYSA-N

General description

5-Bromothiazole is a monobromothiazole, which can be prepared via treatment of 2,5-dibromothiazole with sodium ethoxide followed by hydrogenation over spongy nickel.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

170.1 °F - closed cup

Flash Point(C)

76.7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Synthesis of Brominated Thiazoles via Sequential Bromination-Debromination Methods"
Uzelac.JE and Rasmussen.CS
The Journal of Organic Chemistry (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service