Skip to Content
MilliporeSigma
All Photos(1)

Documents

677140

Sigma-Aldrich

Sodium D-Lactate-3-13C solution

endotoxin tested, 45-55% in H2O ((w/w)), 99 atom % 13C

Synonym(s):

Sodium D-lactate-3-13C solution, D-Lactic -3-13C acid sodium salt solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CH3CH(OH)CO2Na
CAS Number:
Molecular Weight:
113.05
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

description

Packaged as gram of solution, 45-55%(w/w)

isotopic purity

99 atom % 13C

concentration

45-55% in H2O ((w/w))

suitability

endotoxin tested

mass shift

M+1

SMILES string

[Na+].[13CH3][C@@H](O)C([O-])=O

InChI

1S/C3H6O3.Na/c1-2(4)3(5)6;/h2,4H,1H3,(H,5,6);/q;+1/p-1/t2-;/m1./s1/i1+1;

InChI key

NGSFWBMYFKHRBD-MDBPNCOTSA-M

Looking for similar products? Visit Product Comparison Guide

Application

For R&D use only. Not for drug, household, or other uses. Any other use is the sole responsibility of the end user.

Endotoxin testing is done on the bulk form of the product before subdivision packaging. This bulk test does not guarantee that the subdivision or repackaged aliquot is endotoxin-free when it is received or used by the customer and does not imply suitability for any particular purpose. If the product must be endotoxin-free for the intended application, the product should be tested prior to actual use.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service