Sodium D-lactate-3-13C solution
Synonym(s):
D-Lactic -3-13C acid sodium salt solution, Sodium D-lactate-3-13C solution
Linear Formula:
13CH3CH(OH)CO2Na
CAS Number:
Molecular Weight:
113.05
Products
Product Category
Brand
Formula Weight
Isotope
Markush Class
Markush Group
Optical Isomer
Technique
Showing 1-2 of 2