Skip to Content
MilliporeSigma
All Photos(1)

Documents

673145

Sigma-Aldrich

3-Methyl-4-(methylthio)phenol

97%

Synonym(s):

4-(Methylthio)-m-cresol, 3-Methyl-4-(methylthio)phenol, MTMC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10OS
CAS Number:
Molecular Weight:
154.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

56-60 °C

SMILES string

CSc1ccc(O)cc1C

InChI

1S/C8H10OS/c1-6-5-7(9)3-4-8(6)10-2/h3-5,9H,1-2H3

InChI key

VKALYYFVKBXHTF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Chen et al.
IEEE transactions on medical imaging, 37(8), 1775-1787 (2018-07-12)
The O6-methylguanine-DNA methyltransferase (MGMT) promoter methylation and isocitrate dehydrogenase 1 (IDH1) mutation in high-grade gliomas (HGG) have proven to be the two important molecular indicators associated with better prognosis. Traditionally, the statuses of MGMT and IDH1 are obtained via surgical
Youhong Ai et al.
Journal of chromatography. A, 1407, 52-57 (2015-07-15)
A nanostructural polyaniline-poly(propylene oxide) (PANI-PPO) composite coating was electrochemically synthesized on a stainless steel wire, by using acidic ionic liquid 1-sulfobutyl-3-methylimidazolium hydrosulfate as supporting electrolyte. The coating showed strong hydrophobicity and allowed for the direct immersion solid-phase microextraction of carbamate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service