Skip to Content
MilliporeSigma
All Photos(2)

Documents

399353

Sigma-Aldrich

1-Methylbenzimidazole

99%

Synonym(s):

1-Methyl-1H-benzimidazole, 1-Methyl-1H-benzo[d]imidazole, 3-Methylbenzimidazole, N-Methylbenzimidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8N2
CAS Number:
Molecular Weight:
132.16
MDL number:
UNSPSC Code:
12352100
eCl@ss:
32151902
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

154 °C/12 mmHg (lit.)

mp

59-62 °C (lit.)

SMILES string

Cn1cnc2ccccc12

InChI

1S/C8H8N2/c1-10-6-9-7-4-2-3-5-8(7)10/h2-6H,1H3

InChI key

FGYADSCZTQOAFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Methylbenzimidazole is a heterocyclic building block. Effect of the interaction between 1-methylbenzimidazole (MBI) with Li+ and TiO2 on the performance of dye-sensitized solar cells has been investigated.

Application

1-Methylbenzimidazole is the suitable reagent used as electrolyte additive for the dye-sensitized solar cells and solid-state dye-sensitized solar cells.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Kelvin K H Tong et al.
Molecules (Basel, Switzerland), 25(16) (2020-08-17)
Thiones have been investigated as ligands in metal complexes with catalytic and biological activity. We report the synthesis, characterization, and biological evaluation of a series of MII/III complexes of the general formulae [MII(cym)(L)Cl]X (cym = η6-p-cymene) or [MIII(Cp*)(L)Cl]X (Cp* =
Yanyan Fang et al.
Chemistry, an Asian journal, 14(23), 4201-4206 (2019-10-10)
Two types of ionic liquids (ILs), 1-(3-hexenyl)-3-methyl imidazolium iodide and 1-(3-butenyl)-3-methyl imidazolium iodide, are synthesized by introducing an unsaturated bond into the side alkyl chain of the imidazolium cation. These new ionic liquids exhibit high thermal stability and low viscosity
Influence of 1-methylbenzimidazole interactions with Li+ and TiO 2 on the performance of dye-sensitized solar cells.
Zhang C, et al.
Electrochimica Acta, 53(17), 5503-5508 (2008)
Hlaing H Maw et al.
Drug metabolism and disposition: the biological fate of chemicals, 46(6), 770-778 (2018-03-09)
BI 187004, an 11β-hydroxysteroid dehydrogenase 1 inhibitor, was administered once daily for 14 days to eight patients with type 2 diabetes mellitus. N-methylation was identified as a major biotransformation pathway. In four patients treated with BI 187004, the plasma exposure
Optimization of dye-sensitized solar cells prepared by compression method.
Boschloo G, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 148(1), 11-15 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service