Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

660744

Sigma-Aldrich

Palmitic acid-d31

endotoxin tested, 98 atom % D

Synonym(s):

Hexadecanoic-d31 acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3(CD2)14CO2H
CAS Number:
Molecular Weight:
287.62
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

isotopic purity

98 atom % D

form

solid

mp

61-64 °C

suitability

endotoxin tested

mass shift

M+31

SMILES string

[2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(O)=O

InChI

1S/C16H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3,(H,17,18)/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2

InChI key

IPCSVZSSVZVIGE-SAQPIRCFSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

For R&D use only. Not for drug, household, or other uses. Any other use is the sole responsibility of the end user.

Endotoxin testing is done on the bulk form of the product before subdivision packaging. This bulk test does not guarantee that the subdivision or repackaged aliquot is endotoxin-free when it is received or used by the customer and does not imply suitability for any particular purpose. If the product must be endotoxin-free for the intended application, the product should be tested prior to actual use.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhenhua Tian et al.
Frontiers in pharmacology, 11, 910-910 (2020-08-09)
Uncaria rhynchophylla (Miq.) Miq. ex Havil is widely used in the treatment of hypertension. The Uncaria extract and its bioactives, rhynchophylline and isorhynchophylline, reduced the blood pressure and fatty content in liver cells. In the present study, the antihypertensive effects
Qin Yang et al.
Lipids in health and disease, 16(1), 10-10 (2017-01-18)
Dietary polyunsaturated fatty acids (PUFAs), especially n-3 PUFAs, are important for human health. The intestinal tract, a location that is heavily colonized by microorganisms, is the main organ for absorbing fatty acids. The purpose of this study was to analyze
Avarzed Amgalanbaatar et al.
Lipids, 50(8), 799-804 (2015-07-01)
This study demonstrated that the cells of Helicobacter felis and Helicobacter cinaedi spontaneously absorb cholesterol added to the medium. A recent study by our group has revealed that phosphatidylethanolamine (PtdEtn) of Helicobacter pylori contains myristic acid as the most predominant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service