Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

65671

Sigma-Aldrich

(R)-1-{(RP)-2-[2-(Diphenylphosphino)phenyl]ferrocenyl}ethylbis[3,5-bis-(trifluoromethyl)phenyl]phosphine

≥97%

Synonym(s):

(1S)-1-[(1R)-1-[Bis[3,5-bis(trifluoromethyl)phenyl]phosphino]ethyl]-2-[2-(diphenylphosphino)phenyl]ferrocene, Walphos SL-W001-2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H32F12FeP2
CAS Number:
Molecular Weight:
930.52
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97%

optical purity

ee: ≥99%

functional group

fluoro
phosphine

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.C[C@@H]([C]2[CH][CH][CH][C]2c3ccccc3P(c4ccccc4)c5ccccc5)P(c6cc(cc(c6)C(F)(F)F)C(F)(F)F)c7cc(cc(c7)C(F)(F)F)C(F)(F)F

InChI

1S/C41H27F12P2.C5H5.Fe/c1-25(34-16-10-17-35(34)36-15-8-9-18-37(36)55(30-11-4-2-5-12-30)31-13-6-3-7-14-31)54(32-21-26(38(42,43)44)19-27(22-32)39(45,46)47)33-23-28(40(48,49)50)20-29(24-33)41(51,52)53;1-2-4-5-3-1;/h2-25H,1H3;1-5H;/t25-;;/m0../s1

InChI key

LKNKYBWIZNAXBY-WLOLSGMKSA-N

General description

The product is an air-stable and non-hygroscopic Solvias chiral phosphine ligand.

Application

Walphos SL-W001-2, in the presence of a ruthenium catalyst may be used as a ligand in the following processes:
  • Asymmetric hydrogenation of IPCA-D, a cinnamic acid derivative.
  • Addition of 3,5-difluorophenylketimine to 4-methoxyphenylboronic acid to form the corresponding tertiary substituted cyclic sulfamate derivative with high enantioselctivity.2

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

sold in collaboration with Solvias AG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective Rh (I)-catalyzed addition of arylboronic acids to cyclic ketimines.
Kong J, et al.
Organic Letters, 17(22), 5520-5523 (2015)
Afrooz Zirakzadeh et al.
Organometallics, 33(8), 1945-1952 (2014-05-06)
Two representative Walphos analogues with an achiral 2,2″-biferrocenediyl backbone were synthesized. These diphosphine ligands were tested in the rhodium-catalyzed asymmetric hydrogenation of several alkenes and in the ruthenium-catalyzed hydrogenation of two ketones. The results were compared with those previously obtained

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service