Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

656658

Sigma-Aldrich

Ethane-d5 sulfonic acid

98 atom % D, 95% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2D5HO3S
Molecular Weight:
115.16
UNSPSC Code:
12352106
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

95% (CP)

mass shift

M+5

InChI

1S/C2H6O3S/c1-2-6(3,4)5/h2H2,1H3,(H,3,4,5)/i1D3,2D2

InChI key

CCIVGXIOQKPBKL-ZBJDZAJPSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service