Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

56260

Sigma-Aldrich

3-Hydroxypropane-1-sulfonic acid

technical, ≥80% (T)

Synonym(s):

1-Hydroxypropane-3-sulfonic acid, 3-Hydroxy-1-propanesulfonic acid, 3-Hydroxypropan-1-sulfonic acid, 3-Hydroxypropanesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH2CH2SO3H
CAS Number:
Molecular Weight:
140.16
Beilstein:
1753832
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Assay

≥80% (T)

impurities

≤0.1% 1,3-propane sultone
~20% water

density

1.364 g/mL at 20 °C (lit.)

functional group

hydroxyl
sulfonic acid

SMILES string

OCCCS(O)(=O)=O

InChI

1S/C3H8O4S/c4-2-1-3-8(5,6)7/h4H,1-3H2,(H,5,6,7)

InChI key

WQPMYSHJKXVTME-UHFFFAOYSA-N

Related Categories

General description

may contain up to ~40% (NMR) 3,3′-Oxydi(propanesulfonic acid)

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marie Pollard et al.
Journal of psychopharmacology (Oxford, England), 28(10), 935-946 (2014-07-18)
Suppressing anxiety and fear memory relies on bidirectional projections between the medial prefrontal cortex and the amygdala. Positive allosteric modulators of mGluR5 improve cognition in animal models of schizophrenia and retrieval of newly formed associations such as extinction of fear-conditioned

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service