Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

655325

Sigma-Aldrich

Tri-tert-butylphosphine solution

1.0 M in toluene

Synonym(s):

P(t-Bu)3, P(t-Bu)3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H27P
CAS Number:
Molecular Weight:
202.32
MDL number:
UNSPSC Code:
12352128
PubChem Substance ID:
NACRES:
NA.22

Quality Level

reaction suitability

reagent type: ligand
reaction type: Addition Reactions

reagent type: ligand
reaction type: Buchwald-Hartwig Cross Coupling Reaction

reagent type: ligand
reaction type: Heck Reaction

reagent type: ligand
reaction type: Negishi Coupling

reagent type: ligand
reaction type: Sonogashira Coupling

reagent type: ligand
reaction type: Stille Coupling

reagent type: ligand
reaction type: Suzuki-Miyaura Coupling

concentration

1.0 M in toluene

density

0.861 g/mL at 25 °C

functional group

phosphine

SMILES string

CC(C)(C)P(C(C)(C)C)C(C)(C)C

InChI

1S/C12H27P/c1-10(2,3)13(11(4,5)6)12(7,8)9/h1-9H3

InChI key

BWHDROKFUHTORW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Tri-tert-butylphosphine is a suitable ligand for palladium-catalyzed coupling reactions such as:
  • Heck Reactions
  • Negishi cross-coupling
  • Stille reactions
  • Sonogashira reactions

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Corr. 1B - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

39.9 °F

Flash Point(C)

4.4 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pd (PhCN) 2Cl2/P (t-Bu) 3: a versatile catalyst for Sonogashira reactions of aryl bromides at room temperature.
Hundertmark T, et al.
Organic Letters, 2(12), 1729-1731 (2000)
Heck reactions in the presence of P (t-Bu) 3: Expanded scope and milder reaction conditions for the coupling of aryl chlorides.
Littke A F, et al.
The Journal of Organic Chemistry, 64(1), 10-11 (1999)
Pd/P (t-Bu) 3: a mild and general catalyst for Stille reactions of aryl chlorides and aryl bromides
Littke A F, et al.
Journal of the American Chemical Society, 124(22), 6343-6348 (2002)
The first general method for palladium-catalyzed Negishi cross-coupling of aryl and vinyl chlorides: use of commercially available Pd (P (t-Bu) 3) 2 as a catalyst
Dai C, et al.
Journal of the American Chemical Society, 123(12), 2719-2724 (2001)
Gregory C Fu
Accounts of chemical research, 41(11), 1555-1564 (2008-10-25)
Metal-catalyzed coupling reactions of aryl electrophiles with organometallics and with olefins serve as unusually effective tools for forming new carbon-carbon bonds. By 1998, researchers had developed catalysts that achieved reactions of aryl iodides, bromides, and triflates. Nevertheless, many noteworthy challenges

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service