Skip to Content
MilliporeSigma
All Photos(1)

Documents

654426

Sigma-Aldrich

1,7-Dibromoheptane-1,2,6,7-13C4

99 atom % 13C, 97% (CP)

Synonym(s):

Heptamethylene dibromide-1,2,6,7-13C4

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br13CH213CH2(CH2)313CH213CH2Br
CAS Number:
Molecular Weight:
261.96
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

97% (CP)

bp

255 °C (lit.)

density

1.533 g/mL at 25 °C

mass shift

M+4

SMILES string

Br[13CH2][13CH2]CCC[13CH2][13CH2]Br

InChI

1S/C7H14Br2/c8-6-4-2-1-3-5-7-9/h1-7H2/i4+1,5+1,6+1,7+1

InChI key

LVWSZGCVEZRFBT-SQLBHGNYSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2 - Resp. Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service