Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

649899

Sigma-Aldrich

1-(3-Methoxyphenyl)-1-heptanone

97%

Synonym(s):

1-(3-Methoxyphenyl)heptan-1-one, 3-Heptanoylanisole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H20O2
CAS Number:
Molecular Weight:
220.31
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

bp

139-144 °C/0.5 mmHg (lit.)

SMILES string

CCCCCCC(=O)c1cccc(OC)c1

InChI

1S/C14H20O2/c1-3-4-5-6-10-14(15)12-8-7-9-13(11-12)16-2/h7-9,11H,3-6,10H2,1-2H3

InChI key

FIRIFBOVXOXLTG-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service