Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H12455

Sigma-Aldrich

Hexanophenone

99%

Synonym(s):

Amyl phenyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CO(CH2)4CH3
CAS Number:
Molecular Weight:
176.25
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.5105 (lit.)

bp

265 °C (lit.)

mp

25-26 °C (lit.)

density

0.958 g/mL at 25 °C (lit.)

SMILES string

CCCCCC(=O)c1ccccc1

InChI

1S/C12H16O/c1-2-3-5-10-12(13)11-8-6-4-7-9-11/h4,6-9H,2-3,5,10H2,1H3

InChI key

MAHPVQDVMLWUAG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nikola Vaňková et al.
Journal of chromatography. A, 1409, 152-158 (2015-07-29)
The effect of gradient steepness on the kinetic performance limits and peak compression effects has been assessed in gradient mode for the separation of phenol derivatives using columns packed with 2.6μm core-shell particles. The effect of mobile-phase velocity on peak
Martin Gilar et al.
Journal of chromatography. A, 1381, 110-117 (2015-01-22)
Sample introduction in microfluidic liquid chromatography often generates wide zones rather than peaks, especially when a large sample volume (relative to column volume) is injected. Formation of wide injection zones can be further amplified when the sample is dissolved in
Martin Gilar et al.
Journal of chromatography. A, 1390, 86-94 (2015-03-10)
Chromatographic zone broadening is a common issue in microfluidic chromatography, where the sample volume introduced on column often exceeds the column void volume. To better understand the propagation of wide chromatographic zones on a separation device, a series of MS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service