Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

636320

Sigma-Aldrich

2-Amino-5-bromopyrazine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4BrN3
CAS Number:
Molecular Weight:
174.00
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

113-117 °C (lit.)

functional group

bromo

SMILES string

Nc1cnc(Br)cn1

InChI

1S/C4H4BrN3/c5-3-1-8-4(6)2-7-3/h1-2H,(H2,6,8)

InChI key

KRRTXVSBTPCDOS-UHFFFAOYSA-N

Application

Used without prior protection of the amino group in a palladium-catalyzed cross-coupling with pyridylboronic acids leading to pyrazinylpyridines.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amy E Thompson et al.
The Journal of organic chemistry, 70(1), 388-390 (2004-12-31)
A range of halogenated aromatics and heteroaromatics bearing a primary amine group are shown to be suitable substrates for Suzuki cross-coupling reactions with arylboronic acids and pyridylboronic acids under standard conditions, without the need for protection/deprotection steps. New amino-substituted arylpyridines

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service