Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

617407

Sigma-Aldrich

2,6-Difluorobenzamide-α-13C,15N

99 atom % 13C, 98 atom % 15N, 97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H313CO15NH2
CAS Number:
Molecular Weight:
159.10
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

99 atom % 13C
98 atom % 15N

Assay

97% (CP)

form

solid

mp

145-148 °C (lit.)

mass shift

M+2

SMILES string

[15NH2][13C](=O)c1c(F)cccc1F

InChI

1S/C7H5F2NO/c8-4-2-1-3-5(9)6(4)7(10)11/h1-3H,(H2,10,11)/i7+1,10+1

InChI key

AVRQBXVUUXHRMY-ZCIFZXMMSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service