Skip to Content
MilliporeSigma
All Photos(1)

Documents

616125

Sigma-Aldrich

Decanoic-10,10,10-d3 acid

99 atom % D, 99% (CP)

Synonym(s):

Capric acid-10,10,10-d3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3(CH2)8CO2H
CAS Number:
Molecular Weight:
175.28
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

99 atom % D

Quality Level

Assay

99% (CP)

form

solid

bp

268-270 °C (lit.)

mp

30-32 °C (lit.)

density

0.908 g/mL at 25 °C

mass shift

M+3

SMILES string

[2H]C([2H])([2H])CCCCCCCCC(O)=O

InChI

1S/C10H20O2/c1-2-3-4-5-6-7-8-9-10(11)12/h2-9H2,1H3,(H,11,12)/i1D3

InChI key

GHVNFZFCNZKVNT-FIBGUPNXSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pamela Brunswick et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 55(7), 788-799 (2020-04-01)
Observations made for the analysis of the oil spill dispersant tracer dioctyl sulfosuccinate (DOSS) during LC50 toxicity testing, highlighted a stability issue for this tracer compound in seawater. A liquid chromatography high-resolution quadrupole time-of-flight mass spectrometry (LC/QToF) was used to
Pamela Brunswick et al.
Journal of chromatography. A, 1598, 113-121 (2019-04-17)
A high resolution mass spectrometry method was developed for the environmental impact monitoring of oil spill dispersants. Previously reported instability of dioctyl sulfosuccinate (DOSS) dispersant tracer was addressed by the new procedure. The method monitors both DOSS and its degradation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service