Skip to Content
MilliporeSigma
All Photos(1)

Documents

615056

Sigma-Aldrich

Vinyl chloride-d3

≥98 atom % D, ≥99% (CP), contains hydroquinone as stabilizer

Synonym(s):

Chloroethylene-d3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
D2C=CDCl
CAS Number:
Molecular Weight:
65.52
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

≥98 atom % D

Assay

≥99% (CP)

contains

hydroquinone as stabilizer

bp

-13.4 °C (lit.)

mp

-153.8 °C (lit.)

density

0.911 g/mL at 25 °C

mass shift

M+3

SMILES string

[2H]\C([2H])=C(\[2H])Cl

InChI

1S/C2H3Cl/c1-2-3/h2H,1H2/i1D2,2D

InChI key

BZHJMEDXRYGGRV-FUDHJZNOSA-N

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Eye Dam. 1 - Flam. Gas 1A - Muta. 2 - Press. Gas Liquefied gas - Skin Sens. 1

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service