Skip to Content
MilliporeSigma
All Photos(1)

Documents

691976

Sigma-Aldrich

Vinyl-13C2 chloride

stabilized with hydroquinone, 99 atom % 13C, 98% (CP)

Synonym(s):

Chloroethylene-13C2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CH2=13CHCl
CAS Number:
Molecular Weight:
64.48
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

98% (CP)

form

gas

bp

-13.4 °C (lit.)

mp

-153.8 °C (lit.)

mass shift

M+2

SMILES string

Cl[13CH]=[13CH2]

InChI

1S/C2H3Cl/c1-2-3/h2H,1H2/i1+1,2+1

InChI key

BZHJMEDXRYGGRV-ZDOIIHCHSA-N

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Carc. 1A - Eye Irrit. 2 - Flam. Gas 1A - Muta. 2 - Press. Gas Liquefied gas - Skin Sens. 1

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

-77.8 °F - closed cup

Flash Point(C)

-61.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service